Tween® 40

Toimittaja: TCI

T0544-25G T0544-500G
TCIAT0544-25GEA 17.6 EUR
TCIAT0544-25G TCIAT0544-500G
Tween® 40
Tween® 40
Kaava: C₆₂H₁₂₂O₂₆
Kiehumispiste: ? 100 °C (1013 hPa)
Sulamispiste: ∼0,1 °C
Tiheys: 1,05 g/cm³ (20 °C)
Leimahduspiste: ≥ 149 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00165345
CAS-numero: 9005-66-7

Order Now

Testitulosten erittely

Appearance Light yellow to Amber to Dark green clear liquid to cloudy liquid
Water max. 5.0 %
Saponification Value 41.0 to 55.0
Acid Value max. 4.0


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR