Trimethylvinylammonium bromide ≥97.0% (by titrimetric analysis)

Toimittaja: TCI

T0494-1G T0494-10G
TCIAT0494-1GEA 37.4 EUR
TCIAT0494-1G TCIAT0494-10G
Trimethylvinylammonium bromide ≥97.0% (by titrimetric analysis)
Trimethylvinylammonium bromide
Kaava: C₅H₁₂BrN
MDL numero: MFCD00050204
CAS-numero: 10603-92-6
EINECS: 234-227-3

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR