Trifluorietikkahappo ≥99.0% (by titrimetric analysis)

Toimittaja: TCI
Vaara

Synonyms: TFA

T0431-25G T0431-500G T0431-100G
TCIAT0431-25GEA 17.6 EUR
TCIAT0431-25G TCIAT0431-500G TCIAT0431-100G
Trifluorietikkahappo ≥99.0% (by titrimetric analysis)
Trifluorietikkahappo
Kaava: C₂HF₃O₂
Kiehumispiste: 73°C
Sulamispiste: –15 °C
Tiheys: 1,5351 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00004169
CAS-numero: 76-05-1
EINECS: 200-929-3
UN: 2699
ADR: 8,I
Merck Index: 14,09681
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=871b900083dedee58b0800cd3d6f6036f49984a7&vid=198caec32f48ad15322b0355180067e439d6458f&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance Colorless to Almost colorless clear liquid
Purity (Neutralization titration) min. 99.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR