Ethyl-O-toluate ≥98.0% (by GC)

Toimittaja: TCI

T0294-25G T0294-500G
TCIAT0294-25GEA 30.8 EUR
TCIAT0294-25G TCIAT0294-500G
Ethyl-O-toluate ≥98.0% (by GC)
Ethyl-O-toluate
Kaava: C₁₀H₁₂O₂
Kiehumispiste: 227°C
Sulamispiste: –10 °C
Tiheys: 1,0479 g/cm³ (4 °C)
Leimahduspiste: 91°C
MDL numero: MFCD00009112
CAS-numero: 87-24-1
EINECS: 201-734-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=da35a4e1062c0611ad8b33ed1ed8c4d136eba647&vid=da35a4e1062c0611ad8b33ed1ed8c4d136eba647&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR