o-Tolueenihappo ≥98.0% (by titrimetric analysis)

Toimittaja: TCI
Varoitus

T0292-25G T0292-500G T0292-100G
TCIAT0292-25GEA 17.6 EUR
TCIAT0292-25G TCIAT0292-500G TCIAT0292-100G
o-Tolueenihappo ≥98.0% (by titrimetric analysis)
o-Tolueenihappo
Kaava: C₈H₈O₂
Kiehumispiste: 259°C
Sulamispiste: 101…104 °C
Tiheys: 1,2555 g/cm³ (20 °C)
Leimahduspiste: 148 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00002477
CAS-numero: 118-90-1
EINECS: 204-284-9
Merck Index: 14,09535
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=44fba6356c8aa774509fd52ba72f7f9e1cd11814&vid=6756a4eceab66763be8464b58d8862ee92584bbd&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance White to Orange to Green powder to crystal
Purity (Neutralization titration) min. 98.0 %
Melting point 102.0 to 107.0 deg-C
Solubility in Methanol almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR