Ethyl thiocyanate ≥98.0% (by GC)

Toimittaja: TCI
Varoitus

T0200-25ML T0200-500ML
TCIAT0200-25MLEA 183 EUR
TCIAT0200-25ML TCIAT0200-500ML
Ethyl thiocyanate ≥98.0% (by GC)
Ethyl thiocyanate
Kaava: C₃H₅NS
Kiehumispiste: 145°C
Sulamispiste: –86 °C
Tiheys: 1,01 g/cm³ (20 °C)
Leimahduspiste: 42°C
MDL numero: MFCD00001834
CAS-numero: 542-90-5
EINECS: 208-833-3
UN: 1992
ADR: 3,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=61ea27fb839ed49080d046787b98a36e5a879da3&vid=61ea27fb839ed49080d046787b98a36e5a879da3&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR