Tetrabutylammonium hydroxide 10% in isopropanol (IPA)

Toimittaja: TCI
Vaara

T0056-25G T0056-100G
TCIAT0056-25GEA 71.5 EUR
TCIAT0056-25G TCIAT0056-100G
Tetrabutylammonium hydroxide 10% in isopropanol (IPA)
Tetrabutylammonium hydroxide
Kaava: C₁₆H₃₇NO
Storage Temperature: Jääkaappi
MDL numero: MFCD00009425
CAS-numero: 2052-49-5
EINECS: 218-147-6
UN: 2924
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6c6ea4e74328aa9639f6ce5946d2f83848422a43&vid=ad7653994c8cf9b5faf7b48cb7580d2bdfcc9a0d&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance Colorless to Light orange to Yellow clear liquid to slightly cloudy liquid
Concentration (Neutralization titration) 10.0 to 11.0 w/w%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR