Eosiini Y ≥90.0% (by UV/Vis)

Toimittaja: TCI
Varoitus

T0037-25G
TCIAT0037-25GEA 25.3 EUR
TCIAT0037-25G
Eosiini Y ≥90.0% (by UV/Vis)
Eosiini Y
Kaava: C₂₀H₆Br₄Na₂O₅
Sulamispiste: 295…296 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00005040
CAS-numero: 17372-87-1
EINECS: 241-409-6
Merck Index: 14,03603
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=04ac7b35eefa3f959e769f54139597808a2affad&vid=04ac7b35eefa3f959e769f54139597808a2affad&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance Orange to Brown to Dark red powder to crystaline
Purity (UV-vis method) min. 90.0 % (calcd.on dried substance)
Lambda max. 515.0 to 519.0 nm (0.02mol/L AcONH4 sol.)
Drying loss max. 15.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR