(±)-α-Terpineol ≥80.0% (by GC)

Toimittaja: TCI
Varoitus

T0022-25ML T0022-500ML
TCIAT0022-25MLEA 18.7 EUR
TCIAT0022-25ML TCIAT0022-500ML
(±)-α-Terpineol ≥80.0% (by GC)
(±)-α-Terpineol
Kaava: C₁₀H₁₈O
Leimahduspiste: 101°C
MDL numero: MFCD00001557
CAS-numero: 98-55-5
EINECS: 202-680-6
Merck Index: 14,09171
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8a2ae161197eb313a5f7642db02ac8ea0b2557bf&vid=8a2ae161197eb313a5f7642db02ac8ea0b2557bf&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 80.0 %
Specific rotation [a]20/D -15.0 to -40.0 deg (neat)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR