Sodium-2,3,5,6-tetrafluoro-4-hydroxybenzenesulphonate ≥98.0% (by HPLC)

Toimittaja: TCI

S0836-1G S0836-5G
TCIAS0836-1GEA 127 EUR
TCIAS0836-1G TCIAS0836-5G
Sodium-2,3,5,6-tetrafluoro-4-hydroxybenzenesulphonate ≥98.0% (by HPLC)
Sodium-2,3,5,6-tetrafluoro-4-hydroxybenzenesulphonate
Kaava: C₆H₂F₄O₄S
CAS-numero: 221908-17-4

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR