4-Sulphocalix[8]arene hydrate ≥98.0% (by HPLC, titration analysis)

Toimittaja: TCI
Varoitus

S0471-1G S0471-5G
TCIAS0471-1GEA 182 EUR
TCIAS0471-1G TCIAS0471-5G
4-Sulphocalix[8]arene hydrate ≥98.0% (by HPLC, titration analysis)
4-Sulphocalix[8]arene hydrate
Kaava: C₅₆H₄₈O₃₂S₈
MDL numero: MFCD00917032
CAS-numero: 137407-62-6

Order Now

Testitulosten erittely

Appearance White to Gray to Red powder to crystal
Purity (HPLC) min. 98.0 area%
Purity (Neutralization titration) min. 98.0 % (calcd.on anh.substance)
Water max. 20 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR