Monomethyl sebacate ≥98.0% (by titrimetric analysis)

Toimittaja: TCI
Varoitus

S0395-25G S0395-5G
TCIAS0395-25GEA 263 EUR
TCIAS0395-25G TCIAS0395-5G
Monomethyl sebacate ≥98.0% (by titrimetric analysis)
Monomethyl sebacate
Kaava: C₁₁H₂₀O₄
MDL numero: MFCD00004439
CAS-numero: 818-88-2
EINECS: 212-458-0

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (GC) min. 95.0 %
Purity (Neutralization titration) min. 98.0 %
Melting point 40.0 to 44.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR