Cobalt(II) stearate ≥97.0% (by titrimetric analysis)

Toimittaja: TCI
Vaara

S0393-25G
TCIAS0393-25GEA 19.8 EUR
TCIAS0393-25G
Cobalt(II) stearate ≥97.0% (by titrimetric analysis)
Kaava: C₃₆H₇₀CoO₄
Sulamispiste: 75°C
MDL numero: MFCD00054343
CAS-numero: 1002-88-6
EINECS: 213-694-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b6fd8b2779d1552d8632db531e34cc3729545061&vid=b6fd8b2779d1552d8632db531e34cc3729545061&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance Red to Dark blue to Black powder to crystal
Purity (Chelometric Titration) min. 97.0 %
Stearic acid (composition of fatty acid) min. 60.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR