Iron(III) stearate

Toimittaja: TCI

S0256-25G S0256-500G S0256-100G
TCIAS0256-25GEA 30.8 EUR
TCIAS0256-25G TCIAS0256-500G TCIAS0256-100G
Iron(III) stearate
Iron(III) stearate
Kaava: C₁₈H₃₆O₂
MDL numero: MFCD00050716
CAS-numero: 555-36-2
EINECS: 209-095-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6b1a760aec8b5eb2b4872639e605db9d231422fa&vid=6b1a760aec8b5eb2b4872639e605db9d231422fa&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Stearic acid (composition of fatty acid) min. 60.0 %
Iron 5.8 to 7.0 %
Free Acid max. 10.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR