Kaliumsorbaatti ≥99.0% (by titrimetric analysis)

Toimittaja: TCI
Varoitus

S0057-25G S0057-500G
TCIAS0057-25GEA 17.6 EUR
TCIAS0057-25G TCIAS0057-500G
Kaliumsorbaatti ≥99.0% (by titrimetric analysis)
Kaliumsorbaatti
Kaava: CH₃CH=CHCH=CHCOOK
Sulamispiste: 270 °C
Tiheys: 1,36 g/cm³ (20 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00016546
CAS-numero: 24634-61-5
EINECS: 246-376-1
Merck Index: 14,07671
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=76ba039900dd4825054e19c4290f3c8f545693ef&vid=76ba039900dd4825054e19c4290f3c8f545693ef&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance White to Light yellow to Light orange powder to crystal
Purity (Nonaqueous Titration) min. 99.0 %
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR