D(-)-Seriini ≥98.5% (by titrimetric analysis)

Toimittaja: TCI

S0033-25G S0033-5G
TCIAS0033-25GEA 55 EUR
TCIAS0033-25G TCIAS0033-5G
D(-)-Seriini ≥98.5% (by titrimetric analysis)
D(-)-Seriini
Kaava: C₃H₇NO₃
Sulamispiste: 235 °C
MDL numero: MFCD00004269
CAS-numero: 312-84-5
EINECS: 206-229-4
Merck Index: 12,08604
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=dad51a80202c1a334c9909fe58b339861731a9d1&vid=dad51a80202c1a334c9909fe58b339861731a9d1&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (Nonaqueous Titration) min. 98.5 %
Specific rotation [a]20/D -14.3 to -15.5 deg (C=10, HCl (1+5))
Solubility in Water almost transparency

TEKNISET TIEDOT

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR