Diacetato[(R)-(+)-2,2'-bis(di-p-tolylphosphino)-1,1'-binaphthyl]ruthenium(II)

Toimittaja: TCI

R0168-200MG R0168-1G
TCIAR0168-200MGEA 33 EUR
TCIAR0168-200MG TCIAR0168-1G
Diacetato[(R)-(+)-2,2'-bis(di-p-tolylphosphino)-1,1'-binaphthyl]ruthenium(II)
Diacetato[(R)-(+)-2,2'-bis(di-p-tolylphosphino)-1,1'-binaphthyl]ruthenium(II)
MDL numero: MFCD09753021
CAS-numero: 116128-29-1
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d0f3fcbdbdc3d6e9ea84caf85a4afb40f76d610c&vid=d0f3fcbdbdc3d6e9ea84caf85a4afb40f76d610c&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance Dark green to Dark red to Black powder to crystal
IR to pass test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR