Diacetato[(S)-(-)-2,2'-bis(diphenylphosphino)-1,1'-binaphthyl]ruthenium(II)

Toimittaja: TCI

R0167-200MG R0167-1G
TCIAR0167-200MGEA 40.7 EUR
TCIAR0167-200MG TCIAR0167-1G
Diacetato[(S)-(-)-2,2'-bis(diphenylphosphino)-1,1'-binaphthyl]ruthenium(II)
Diacetato[(S)-(-)-2,2'-bis(diphenylphosphino)-1,1'-binaphthyl]ruthenium(II)
Kaava: C₄₈H₃₈O₄P₂Ru
MDL numero: MFCD09753020
CAS-numero: 261948-85-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5d99681fb817458461c388a179a88901a439b8f2&vid=5d99681fb817458461c388a179a88901a439b8f2&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance Yellow to Amber to Dark purple powder to crystal
IR to pass test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR