[RuCl(p-cymene)((S)-segphos®)]Cl

Toimittaja: TCI

R0155-200MG R0155-1G
TCIAR0155-200MGEA 36.3 EUR
TCIAR0155-200MG TCIAR0155-1G
[RuCl(p-cymene)((S)-segphos®)]Cl
[RuCl(p-cymene)((S)-segphos®)]Cl
Kaava: C₄₈H₄₂Cl₂O₄P₂Ru
MDL numero: MFCD09753017
CAS-numero: 944451-29-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f98932c8df12177777dd342acad7327a0532b56f&vid=f98932c8df12177777dd342acad7327a0532b56f&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance Light yellow to Brown to Dark green powder to crystal
IR to pass test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR