Ru-MACHO®-BH

Toimittaja: TCI
Vaara

R0137-200MG R0137-1G
TCIAR0137-200MGEA 38.5 EUR
TCIAR0137-200MG TCIAR0137-1G
Ru-MACHO®-BH
Ru-MACHO®-BH
Kaava: C₂₉H₃₄BNOP₂Ru
MDL numero: MFCD20922901
CAS-numero: 1295649-41-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5c6e33ae1ca2aa9781cfd080df133858ad29e625&vid=5c6e33ae1ca2aa9781cfd080df133858ad29e625&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance White to Yellow to Orange powder to crystal
Content (Ruthenium) 15 to 17 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR