RuCl2[(R)-xylbinap][(R)-daipen]

Toimittaja: TCI

R0133-200MG R0133-1G
TCIAR0133-200MGEA 48.4 EUR
TCIAR0133-200MG TCIAR0133-1G
RuCl2[(R)-xylbinap][(R)-daipen]
RuCl2[(R)-xylbinap][(R)-daipen]
Kaava: C₇₁H₇₄Cl₂N₂O₂P₂Ru
MDL numero: MFCD09753026
CAS-numero: 220114-32-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=31f795baa764f56a74c80169ea83b87790fdfbdd&vid=31f795baa764f56a74c80169ea83b87790fdfbdd&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance Light yellow to Amber to Dark green powder to crystal
IR to pass test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR