Rocuronium bromide ≥90.0% (by titrimetric analysis)

Toimittaja: TCI
Varoitus

R0099-100MG
TCIAR0099-100MGEA 91.3 EUR
TCIAR0099-100MG
Rocuronium bromide ≥90.0% (by titrimetric analysis)
Rocuronium bromide
Kaava: C₃₂H₅₃BrN₂O₄
Sulamispiste: 169°C
Storage Temperature: Jääkaappi
MDL numero: MFCD00867768
CAS-numero: 119302-91-9
Merck Index: 14,08246
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=93d65b8278d95a0373a898d306ab2342c96f4bd6&vid=93d65b8278d95a0373a898d306ab2342c96f4bd6&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance White to Light yellow to Light orange powder to crystal
Purity (Nonaqueous Titration) min. 90.0 %
Specific rotation [a]20/D +28.5 to +32.0 deg (C=1, 0.05mol/L HCl) (calcd.on anh.substance)
Water max. 6.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR