ɑ-D-Ribose-1,5-bis(phosphate) tetrasodium salt

Toimittaja: TCI

R0082-5MG
TCIAR0082-5MGEA 487 EUR
TCIAR0082-5MG
ɑ-D-Ribose-1,5-bis(phosphate) tetrasodium salt
alpha-D-Ribose-1,5-bis(phosphate) tetrasodium salt
Kaava: C₅H₁₂O₁₁P₂
Storage Temperature: Pakastin
CAS-numero: 113599-17-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=25d027cfe92e55c37387fc46071ed382d5194857&vid=25d027cfe92e55c37387fc46071ed382d5194857&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (HPLC) (CAD) min. 98.0 area%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR