Rose Bengal

Toimittaja: TCI

R0041-25G
TCIAR0041-25GEA 47.3 EUR
TCIAR0041-25G
Rose Bengal
Rose Bengal
Kaava: C₂₀H₂Cl₄I₄O₅Na₂
MDL numero: MFCD00151169
CAS-numero: 632-69-9
EINECS: 211-183-3

Order Now

Testitulosten erittely

Appearance Dark red to Brown powder to crystal
Lambda max. 547.0 to 551.0 nm (0.02mol/L AcONH4 sol.)
Absorbance (E1%1cm) min. 900 (0.02mol/L AcONH4 sol., 547.0 to 551.0 nm)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR