Quinidine sulphate dihydrate ≥98.0% (by titrimetric analysis)

Toimittaja: TCI
Vaara

Q0010-25G Q0010-5G
TCIAQ0010-25GEA 132 EUR
TCIAQ0010-25G TCIAQ0010-5G
Quinidine sulphate dihydrate ≥98.0% (by titrimetric analysis)
Quinidine sulphate dihydrate
Kaava: C₄₀H₄₈N₄O₄.H₂O₄S·2H₂O
MDL numero: MFCD00149346
CAS-numero: 6591-63-5
UN: 2811
ADR: 6.1,III
Merck Index: 14,08060

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (HPLC) min. 80.0 area%
Purity (Neutralization titration) min. 98.0 % (calcd.on anh.substance)
Specific rotation [a]20/D +268 to +280 deg (C=2, 0.1mol/L HCl)
Water 4.0 to 5.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR