2,2'-[1,4-Phenylenebis(oxy-2,1-ethanediyloxy)]diethanol ≥98.0% (by GC)

Toimittaja: TCI

P2762-1G P2762-5G
TCIAP2762-1GEA 97.9 EUR
TCIAP2762-1G TCIAP2762-5G
2,2'-[1,4-Phenylenebis(oxy-2,1-ethanediyloxy)]diethanol ≥98.0% (by GC)
2,2'-[1,4-Phenylenebis(oxy-2,1-ethanediyloxy)]diethanol
Kaava: C₁₄H₂₂O₆
MDL numero: MFCD01109251
CAS-numero: 35648-87-4

Order Now

Testitulosten erittely

Appearance White to Light yellow to Dark green powder to crystal
Purity (GC) min. 98.0 %
Melting point 76.0 to 80.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR