Polyethylene glycol diacrylate (PEGDA) n=∼4 stabiloitu

Toimittaja: TCI
Vaara


Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

P2756-25G P2756-500G
TCIAP2756-25GEA 20.9 EUR
TCIAP2756-25G TCIAP2756-500G
Polyethylene glycol diacrylate (PEGDA) n=∼4 stabiloitu
Polyethylene glycol diacrylate (PEGDA)
Kaava: CH₂=COC(OCH₂CH₂)nOCOC=CH₂
Leimahduspiste: 104°C
Storage Temperature: Jääkaappi
MDL numero: MFCD00081876
CAS-numero: 26570-48-9

Order Now

Testitulosten erittely

Appearance Colorless to Almost colorless clear liquid
The average value of n 3.5 to 4.5


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR