Polyethylene glycol monododecyl ether n=∼23

Toimittaja: TCI
Varoitus

P2737-25G P2737-500G
TCIAP2737-25GEA 17.6 EUR
TCIAP2737-25G TCIAP2737-500G
Polyethylene glycol monododecyl ether n=∼23
Polyethylene glycol monododecyl ether
Kaava: HO(CH₂CH₂O)nC₁₂H₂₅
MDL numero: MFCD00080891
CAS-numero: 9002-92-0
EINECS: 500-002-6
UN: 3077
ADR: 9,III
Merck Index: 14,07558

Order Now

Testitulosten erittely

Appearance White to Almost white powder to lump
Freezing point 33.0 to 38.0 deg-C
The average value of n 21.0 to 25.0


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR