Polyethylene glycol methyl ether acrylate n=∼9 stabiloitu

Toimittaja: TCI


Stabilization: Stabilised with 4-methoxyphenol (MEHQ)

P2698-25G P2698-500G
TCIAP2698-25GEA 37.4 EUR
TCIAP2698-25G TCIAP2698-500G
Polyethylene glycol methyl ether acrylate n=∼9 stabiloitu
Polyethylene glycol methyl ether acrylate
Kaava: CH₃(OCH₂CH₂)nOCOC=CH₂
MDL numero: MFCD00803685
CAS-numero: 32171-39-4

Order Now

Testitulosten erittely

Appearance Colorless to Light yellow clear liquid
The average value of n 8.5 to 9.5


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR