Pyrimidine-2-sulphonyl fluoride ≥98.0% (by GC)

Toimittaja: TCI
Vaara

P2465-1G P2465-5G
TCIAP2465-1GEA 118 EUR
TCIAP2465-1G TCIAP2465-5G
Pyrimidine-2-sulphonyl fluoride ≥98.0% (by GC)
Pyrimidine-2-sulphonyl fluoride
Kaava: C₄H₃FN₂O₂S
Storage Temperature: Jääkaappi
MDL numero: MFCD09265503
CAS-numero: 35762-87-9
UN: 3261
ADR: 8,II

Order Now

Testitulosten erittely

Purity (GC) min. 98.0 %
Purity (Neutralization back titration) min. 97.0 %
Melting point 58.0 to 62.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR