1-Propyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole ≥98.0% (by GC, titration analysis)

Toimittaja: TCI
Varoitus

P2277-1G P2277-5G
TCIAP2277-1GEA 51.7 EUR
TCIAP2277-1G TCIAP2277-5G
1-Propyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole ≥98.0% (by GC, titration analysis)
1-Propyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazole
Kaava: C₁₂H₂₁BN₂O₂
Kiehumispiste: 102°C
MDL numero: MFCD05663871
CAS-numero: 827614-69-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0c078c5e5b0c85b1a29ffe2007013bbe9adefcdf&vid=0c078c5e5b0c85b1a29ffe2007013bbe9adefcdf&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR