1-[(2-Propynyloxy)methyl]pyrene ≥98.0% (by GC)

Toimittaja: TCI
Varoitus

P2226-200MG P2226-1G
TCIAP2226-200MGEA 213 EUR
TCIAP2226-200MG TCIAP2226-1G
1-[(2-Propynyloxy)methyl]pyrene ≥98.0% (by GC)
1-[(2-Propynyloxy)methyl]pyrene
Kaava: C₂₀H₁₄O
MDL numero: MFCD28384153
CAS-numero: 1115084-83-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e4a184360d8fa70e2d3f985abbe4d528ac58d266&vid=e4a184360d8fa70e2d3f985abbe4d528ac58d266&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 83.0 to 87.0 deg-C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR