Polyethylene glycol monomethyl ether 1000

Toimittaja: TCI

P2185-100G P2185-500G
TCIAP2185-100GEA 26.4 EUR
TCIAP2185-100G TCIAP2185-500G
Polyethylene glycol monomethyl ether 1000
Polyethylene glycol monomethyl ether
Kaava: CH₃(OCH₂CH₂)nOH
MDL numero: MFCD00084416
CAS-numero: 9004-74-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4f61a8b01034308604da1b61335d4e12b444c73f&vid=4f61a8b01034308604da1b61335d4e12b444c73f&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance White to Almost white powder to lump
Average molecular weight 950 to 1,050
Hydroxyl Value 50.0 to 60.0


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR