(3-Phenyl-1H-inden-1-ylidene)bis(tricyclohexylphosphine)ruthenium(II) dichloride tetrahydrofuran ≥97.0% (by titrimetric analysis)

Toimittaja: TCI
Varoitus

P2004-1G
TCIAP2004-1GEA 323 EUR
TCIAP2004-1G
(3-Phenyl-1H-inden-1-ylidene)bis(tricyclohexylphosphine)ruthenium(II) dichloride tetrahydrofuran ≥97.0% (by titrimetric analysis)
(3-Phenyl-1H-inden-1-ylidene)bis(tricyclohexylphosphine)ruthenium(II) dichloride tetrahydrofuran
Kaava: C₅₁H₇₆Cl₂P₂Ru
MDL numero: MFCD07369034
CAS-numero: 250220-36-1
EINECS: 480-140-0

Order Now

Testitulosten erittely

Appearance Amber to Brown powder to crystal
Purity (Argentmetric Titration) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR