Palladium 5% on barium carbonate

Toimittaja: TCI
Vaara

P1701-10G
TCIAP1701-10GEA 77 EUR
TCIAP1701-10G
Palladium 5% on barium carbonate
Palladium
Kaava: Pd
MDL numero: MFCD03427451
CAS-numero: 7440-05-3
EINECS: 231-115-6
UN: 1564
ADR: 6.1,III
Merck Index: 14,06989
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=108d92c667ad8f10b0db60d7aac3c4f2f50f8b4e&vid=e0f05e583835ae6f09a369e777d22dc0ecf6fb3b&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Content (Palladium) 4.50 to 5.50 %
Water max. 3.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR