3-Pyridineboronic acid (contains varying amounts of Anhydride)

Toimittaja: TCI
Varoitus

P1673-1G P1673-25G P1673-5G
TCIAP1673-1GEA 38.5 EUR
TCIAP1673-1G TCIAP1673-25G TCIAP1673-5G
3-Pyridineboronic acid (contains varying amounts of Anhydride)
3-Pyridineboronic acid
Kaava: C₅H₆BNO₂
Storage Temperature: Jääkaappi
MDL numero: MFCD00674177
CAS-numero: 1692-25-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=72112dab2471ed995e1b463122a655f959f9785f&vid=72112dab2471ed995e1b463122a655f959f9785f&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance White to Light yellow powder to crystal
Purity (Neutralization titration) 97.0 to 117.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR