Trimethoxy[3-(phenylamino)propyl]silane ≥98.0% (by titrimetric analysis)

Toimittaja: TCI
Vaara

P1458-25G P1458-250G
TCIAP1458-25GEA 26.4 EUR
TCIAP1458-25G TCIAP1458-250G
Trimethoxy[3-(phenylamino)propyl]silane ≥98.0% (by titrimetric analysis)
Trimethoxy[3-(phenylamino)propyl]silane
Kaava: C₁₂H₂₁NO₃Si
Kiehumispiste: 310°C
Leimahduspiste: 165°C
MDL numero: MFCD00053673
CAS-numero: 3068-76-6
EINECS: 221-328-2
UN: 3267
ADR: 8,III

Order Now

Testitulosten erittely

Appearance Colorless to Light yellow to Light orange clear liquid
Purity (GC) min. 97.0 %
Purity (Nonaqueous Titration) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR