Tris(2,4-di-tert-butylphenyl)phosphite ≥98.0% (by GC)

Toimittaja: TCI
Varoitus

P1421-25G P1421-500G P1421-100G
TCIAP1421-25GEA 18.7 EUR
TCIAP1421-25G TCIAP1421-500G TCIAP1421-100G
Tris(2,4-di-tert-butylphenyl)phosphite ≥98.0% (by GC)
Tris(2,4-di-tert-butylphenyl)phosphite
Kaava: C₄₂H₆₃O₃P
MDL numero: MFCD00072706
CAS-numero: 31570-04-4
EINECS: 250-709-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e4ef987b7e7666437e4344d0946801e59d277fd6&vid=e4ef987b7e7666437e4344d0946801e59d277fd6&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (GC) min. 98.0 %
Melting point 185.0 to 189.0 deg-C
Solubility in Toluene almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR