Methyl pentafluorobenzoate ≥98.0% (by GC)

Toimittaja: TCI
Varoitus

P1418-25G P1418-5G
TCIAP1418-25GEA 110 EUR
TCIAP1418-25G TCIAP1418-5G
Methyl pentafluorobenzoate ≥98.0% (by GC)
Methyl pentafluorobenzoate
Kaava: C₈H₃F₅O₂
Leimahduspiste: 78°C
MDL numero: MFCD00012172
CAS-numero: 36629-42-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f253072b4b80bc30af38bbeee265d614d29a12df&vid=f253072b4b80bc30af38bbeee265d614d29a12df&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR