Sykloheksen-1-yylibentseeni ≥95.0% (by GC)

Toimittaja: TCI

P1302-25ML P1302-5ML
TCIAP1302-25MLEA 130 EUR
TCIAP1302-25ML TCIAP1302-5ML
Sykloheksen-1-yylibentseeni ≥95.0% (by GC)
Sykloheksen-1-yylibentseeni
Kaava: C₁₂H₁₄
Kiehumispiste: 252°C
Sulamispiste: 0 °C
Tiheys: 1,004 g/cm³ (0 °C)
Leimahduspiste: 103°C
MDL numero: MFCD00001542
CAS-numero: 771-98-2
EINECS: 212-242-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3d966d766c789bf4335f8e194ccb34b7733a579c&vid=18b20d2b83fc9bd01b1f1084a9de36a9824ad88b&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 95.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR