Dibenzyl dicarbonate ≥97.0% (by titrimetric analysis)

Toimittaja: TCI
Varoitus

P1281-25G P1281-5G
TCIAP1281-25GEA 365 EUR
TCIAP1281-25G TCIAP1281-5G
Dibenzyl dicarbonate ≥97.0% (by titrimetric analysis)
Dibenzyl dicarbonate
Kaava: C₁₆H₁₄O₅
Storage Temperature: Pakastin
MDL numero: MFCD00043124
CAS-numero: 31139-36-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=044c13831c21cfddbdbccf1e01a9ee643ad4a2f9&vid=044c13831c21cfddbdbccf1e01a9ee643ad4a2f9&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (Morpholine Method) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR