Pyridinium-p-toluenesulphonate ≥98.0% (by titrimetric analysis)

Toimittaja: TCI
Varoitus

P0942-25G P0942-500G P0942-100G
TCIAP0942-25GEA 30.8 EUR
TCIAP0942-25G TCIAP0942-500G TCIAP0942-100G
Pyridinium-p-toluenesulphonate ≥98.0% (by titrimetric analysis)
Pyridinium-p-toluenesulphonate
Kaava: C₁₂H₁₃NO₃S
MDL numero: MFCD00013108
CAS-numero: 24057-28-1
EINECS: 246-002-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d6641d8938b173133dbd0c51aab0a2a56b1e575f&vid=d6641d8938b173133dbd0c51aab0a2a56b1e575f&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystal
Purity (Neutralization titration) min. 98.0 %
Melting point 118.0 to 122.0 deg-C
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR