Polyethylene glycol mono-4-octylphenyl ether n=∼10

Toimittaja: TCI
Varoitus

P0873-25G P0873-500G
TCIAP0873-25GEA 18.7 EUR
TCIAP0873-25G TCIAP0873-500G
Polyethylene glycol mono-4-octylphenyl ether n=∼10
Polyethylene glycol mono-4-octylphenyl ether
Kaava: C₈H₁₇C₆H₄(OCH₂CH₂)nOH
MDL numero: MFCD00132505
CAS-numero: 9002-93-1
Merck Index: 14,06761
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ce22e6aa971860788d1735b7365f04a348dc6f74&vid=32cecd182803afa2348ef9fe0b920ffa91ac0123&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance Colorless to Light yellow clear liquid
The average value of n 8.5 to 11.5


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR