Methyl pentadecanoate ≥98.0% (by GC)

Toimittaja: TCI

P0869-25ML P0869-5ML
TCIAP0869-25MLEA 286 EUR
TCIAP0869-25ML TCIAP0869-5ML
Methyl pentadecanoate ≥98.0% (by GC)
Methyl pentadecanoate
Kaava: C₁₆H₃₂O₂
Kiehumispiste: 155°C
MDL numero: MFCD00008989
CAS-numero: 7132-64-1
EINECS: 230-430-6

Order Now

Testitulosten erittely

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR