Dichloro(methyl)(phenyl)silane ≥98.0% (by GC)

Toimittaja: TCI
Vaara

P0790-25G P0790-250G
TCIAP0790-25GEA 41.8 EUR
TCIAP0790-25G TCIAP0790-250G
Dichloro(methyl)(phenyl)silane ≥98.0% (by GC)
Dichloro(methyl)(phenyl)silane
Kaava: C₇H₈Cl₂Si
Kiehumispiste: 205°C
Leimahduspiste: 82°C
MDL numero: MFCD00000490
CAS-numero: 149-74-6
EINECS: 205-746-2
UN: 2437
ADR: 8,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b289c5b7768fd71344701714a3b7bb1eaf6d552f&vid=b289c5b7768fd71344701714a3b7bb1eaf6d552f&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR