Polyethylene glycol stearylamine n=∼10

Toimittaja: TCI

P0724-500G P0724-25G
TCIAP0724-500GEA 50.6 EUR
TCIAP0724-500G TCIAP0724-25G
Polyethylene glycol stearylamine n=∼10
Polyethylene glycol stearylamine

Order Now

Testitulosten erittely

Appearance Red to Dark red to Brown clear liquid
Purity (Neutralization titration) min. 90.0 %
The average value of n 8.0 to 12.0


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR