Polyethylene glycol monostearate (palmitate and stearate mixture) n=∼40

Toimittaja: TCI

P0721-25G P0721-500G
TCIAP0721-25GEA 17.6 EUR
TCIAP0721-25G TCIAP0721-500G
Polyethylene glycol monostearate (palmitate and stearate mixture) n=∼40
Polyethylene glycol monostearate
Kaava: HO(CH₂CH₂O)nOCC₁₇H₃₅
MDL numero: MFCD00148007
CAS-numero: 9004-99-3

Order Now

Testitulosten erittely

Appearance White to Almost white wax
The average value of n 35.0 to 47.0


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR