2-Pentyyni ≥97.0% (by GC)

Toimittaja: TCI
Vaara

P0628-25ML P0628-5ML
TCIAP0628-25MLEA 156 EUR
TCIAP0628-25ML TCIAP0628-5ML
2-Pentyyni ≥97.0% (by GC)
2-Pentyyni
Kaava: C₅H₈
Kiehumispiste: 56°C
Sulamispiste: –109 °C
Tiheys: 0,7122 g/cm³ (20 °C)
Leimahduspiste: -20°C
Storage Temperature: Jääkaappi
MDL numero: MFCD00009380
CAS-numero: 627-21-4
EINECS: 210-989-2
UN: 3295
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f8a04ee63cad57f1c6e0b53647f2f0049a8f9e34&vid=d12960b6b5e543a1fa04314b23ce18a34d40f199&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance Colorless to Light orange to Yellow clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR