Pyridiini-2,6-dikarboksyylihappo ≥98.0% (by GC, titration analysis)

Toimittaja: TCI
Varoitus

P0554-25G P0554-500G P0554-100G
TCIAP0554-25GEA 22 EUR
TCIAP0554-25G TCIAP0554-500G TCIAP0554-100G
Pyridiini-2,6-dikarboksyylihappo ≥98.0% (by GC, titration analysis)
Pyridiini-2,6-dikarboksyylihappo
Kaava: C₇H₅NO₄
Kiehumispiste: 357 °C (1013 hPa)
Sulamispiste: 252°C
Leimahduspiste: 188 °C
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00006299
CAS-numero: 499-83-2
EINECS: 207-894-3
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=113734767f845c8e82fbb084f4189bf2c50631f2&vid=113734767f845c8e82fbb084f4189bf2c50631f2&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance White to Almost white powder to crystaline
Purity (GC) min. 98.0 %
Purity (Neutralization titration) min. 98.0 %
Solubility in hot Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR