L(-)-proliini ≥99.0% (by HPLC, titration analysis)

Toimittaja: TCI

P0481-25G P0481-250G
TCIAP0481-25GEA 23.1 EUR
TCIAP0481-25G TCIAP0481-250G
L(-)-proliini ≥99.0% (by HPLC, titration analysis)
L(-)-proliini
Kaava: C₅H₉NO₂
Kiehumispiste: 245 °C (1013 hPa)
Sulamispiste: 220…222 °C
Tiheys: 1,35 g/cm³ (25 °C)
Storage Temperature: Huoneenlämpö
MDL numero: MFCD00064318
CAS-numero: 147-85-3
EINECS: 205-702-2
Merck Index: 14,07780
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b40e836604d92b1c16dfcfacf93fd34e26cb8eac&vid=b40e836604d92b1c16dfcfacf93fd34e26cb8eac&prid=web&clid=SAPDAM

Order Now

Testitulosten erittely

Appearance White powder to crystal
Purity (HPLC) (CAD) min. 99.0 area%
Purity (Nonaqueous Titration) min. 99.0 %
Specific rotation [a]20/D -84.0 to -86.0 deg (C=4, H2O)
Solubility in Water almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Lue lisää About VWR